missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Psoralen, MedChemExpress
Description
Psoralen (Ficusin) is a coumarin isolated from the seeds of Fructus Psoraleae. Psoralen exhibits a wide range of biological properties, including anti-cancer, antioxidant, antidepressant, anticancer, antibacterial, and antiviral, et al.
Specifications
Specifications
| Chemical Name or Material | Psoralen |
| Physical Form | Solid |
| CAS | 66-97-7 |
| Color | Off-White |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C11H6O3 |
| For Use With (Application) | Cancer-programmed cell death |
| Synonym | Ficusin |
| Solubility Information | DMSO : 100 mg/mL (537.17 mM; Need ultrasonic) ∣H2O : 1 mg/mL (5.37 mM; ultrasonic and warming and heat to 80°C) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?