missing translation for 'onlineSavingsMsg'
Learn More

Psoralidin, MedChemExpress

Product Code. 16461348
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16461348 5 mg 5mg
16438908 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16461348 Supplier Medchem Express Supplier No. HYN0232

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Psoralidin is a dual inhibitor of COX-2 and 5-LOX, regulates ionizing radiation (IR)-induced pulmonary inflammation.Anti-cancer, anti-bacterial, and anti-inflammatory properties. Psoralidin significantly downregulates NOTCH1 signaling. Psoralidin also greatly induces ROS generation.

Psoralidin is a dual inhibitor of COX-2 and 5-LOX, regulates ionizing radiation (IR)-induced pulmonary inflammation.Anti-cancer, anti-bacterial, and anti-inflammatory properties. Psoralidin significantly downregulates NOTCH1 signaling. Psoralidin also greatly induces ROS generation.

Specifica

Chemical Name or Material Psoralidin
Physical Form Solid
CAS 18642-23-4
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C20H16O5
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 50 mg/mL (148.66 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C1C2=C(OC3=CC(O)=CC=C32)C4=CC(C/C=C(C)\C)=C(O)C=C4O1
Molecular Weight (g/mol) 336.34
Formula Weight 336.34
Percent Purity 98.13%
Grade Research
Vedi altri risultati Mostra meno risultati

Research purposes only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.