missing translation for 'onlineSavingsMsg'
Learn More

PSP/BPIFA2 Antibody (4C7D7) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

203.00 € - 552.00 €

Specifications

Antigen PSP/BPIFA2
Concentration 1 mg/mL
Dilution Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready
Applications Western Blot, Flow Cytometry, ELISA, CyTOF
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18179767
View Documents
Novus Biologicals
NBP2-37438
0.1 mL
552.00 €
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18615225
View Documents
Novus Biologicals
NBP2-37438-0.025ml
0.025 mL
203.00 €
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PSP/BPIFA2 Monoclonal specifically detects PSP/BPIFA2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.
Specifications

Specifications

PSP/BPIFA2
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready
Monoclonal
Mouse
Human
bA49G10.1, chromosome 20 open reading frame 70, Parotid secretory protein, PSP, short palate, lung and nasal epithelium carcinoma associated 2, short palate, lung and nasal epithelium carcinoma-associated protein 2, SPLUNC2
BPIFA2
IgG1
Affinity Purified
1 mg/mL
Western Blot, Flow Cytometry, ELISA, CyTOF
Unconjugated
RUO
PBS with 0.05% Sodium Azide
140683
Purified recombinant fragment of human Splunc2 (AA: 16-250) expressed in E. Coli.
Primary
27 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.