missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTBP2 Antibody (SR1976), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22638-50ul
This item is not returnable.
View return policy
Description
PTBP2 Monoclonal antibody specifically detects PTBP2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| PTBP2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| SR1976 | |
| Western Blot 1:500-1:2000 | |
| brPTB, FLJ34897, neural polypyrimidine tract binding protein, Neural polypyrimidine tract-binding protein, Neurally-enriched homolog of PTB, NPTB, nPTB6, nPTB7, nPTB8, polypyrimidine tract binding protein 2, polypyrimidine tract-binding protein 2, PTB-like protein, PTBPTBLPnPTB5, splicing regulator | |
| A synthesized peptide derived from human PTBP2 (Uniprot #: Q9UKA9) | |
| 50 μg | |
| Neuroscience | |
| 58155 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction