Learn More
PTCD3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA559455
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: PATSLNCIAI LFLRAGRTQE AWKMLGLFRK HNKIPRSELL NELMDSAKVS NSPSQAIEVV ELASAFSLPI CEGLTQRVMS DFAINQEQKE ALSNLTAL Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 74%.
Mitochondrial RNA-binding protein that has a role in mitochondrial translation.Specifications
PTCD3 | |
Polyclonal | |
Unconjugated | |
PTCD3 | |
2610034F17Rik; 2810422B04Rik; 28S ribosomal protein S39, mitochondrial; AA589622; AU045708; Mitochondrial small ribosomal subunit protein mS39; Mrps39; MRP-S39; pentatricopeptide repeat domain 3; pentatricopeptide repeat domain-containing protein 3, mitochondrial; pentatricopeptide repeat-containing protein 3, mitochondrial; Ptcd3; similar to CG4679-PA; transformation-related gene 15 protein; TRG15; TRG-15 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55037 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.8 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96EY7 | |
PTCD3 | |
Recombinant protein corresponding to Human PTCD3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only