missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTOV1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93875-0.1ml
This item is not returnable.
View return policy
Description
PTOV1 Polyclonal antibody specifically detects PTOV1 in Human samples. It is validated for Western Blot
Specifications
| PTOV1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| ACID2, Activator interaction domain-containing protein 2, DKFZp586I111, MGC71475, prostate tumor overexpressed 1, prostate tumor overexpressed gene 1, PTOV-1 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human PTOV1 (NP_001292037.1). MRSPAVPTPARGQLGVAFVLLPPHSEGARVFGALGPIGPSSPGLTLGGLAVSEHRLSNKLLAWSGVLEWQEKRRPYSDSTAKLKRTLPCQAYVNQGENLE | |
| 0.1 mL | |
| Cancer, Cell Biology, Cell Cycle and Replication | |
| 53635 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction