missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTP alpha/PTPRA Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00 € - 550.00 €
Specifications
| Antigen | PTP alpha/PTPRA |
|---|---|
| Dilution | Western Blot 1:1000 - 1:5000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30232260
|
Novus Biologicals
NBP3-35238-100ul |
100 μL |
550.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30227586
|
Novus Biologicals
NBP3-35238-20ul |
20 μL |
190.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PTP alpha/PTPRA Polyclonal antibody specifically detects PTP alpha/PTPRA in Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| PTP alpha/PTPRA | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Phospho Specific, Protein Phosphatase, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 5786 | |
| IgG | |
| Affinity purified |
| Western Blot 1:1000 - 1:5000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| EC 3.1.3.48, HEPTP, LRPHPTPalpha, protein tyrosine phosphatase, receptor type, A, Protein-tyrosine phosphatase alpha, PTPAreceptor type, alpha polypeptide, PTPLCA-related phosphatase, receptor-type tyrosine-protein phosphatase alpha, RPTPA, R-PTP-alpha, tyrosine phosphatase alpha | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 175-295 of human PTP alpha/PTPRA (NP_543030.1).,, Sequence:, GSHSNSFRLSNGRTEDVEPQSVPLLARSPSTNRKYPPLPVDKLEEEINRRMADDNKLFREEFNALPACPIQATCEAASKEENKEKNRYVNILPYDHSRVHLTPVEGVPDSDYINASFINGY | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title