Learn More
PTP1B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11334-1-AP-150UL
 View more versions of this product
Product Code. 16878333
  • 410.00 € / 150µL
Description
Protects DRG1 from proteolytic degradation.
Specifications
Specifications
PTP1B
Polyclonal
Unconjugated
PTPN1
PTP 1B, PTP1B, PTPN1
Rabbit
Antigen Affinity Chromatography
RUO
19246, 24697, 5770
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.35 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P18031, P20417, P35821
PTPN1
PTP1B Fusion Protein Ag1878
150 μL
Primary
Human, Mouse, Rat