missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTPRK Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-30977
This item is not returnable.
View return policy
Description
PTPRK Polyclonal specifically detects PTPRK in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| PTPRK | |
| Polyclonal | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Q15262 | |
| PTPRK | |
| This antibody was developed against a recombinant protein corresponding to amino acids: MTHMVNAMDRSYADQSTLHAEDPLSITFMDQHNFSPRYENHSATAESSRLLDVPRYLCE | |
| 0.1 mL | |
| Protein Phosphatase | |
| 5796 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| DKFZp686C2268, DKFZp779N1045, EC 3.1.3.48, protein tyrosine phosphatase kappa protein tyrosine phosphatase kappa, protein tyrosine phosphatase, receptor type, K, Protein-tyrosine phosphatase kappa, protein-tyrosine phosphatase, receptor type, kappa, PTPK, receptor type, K (R-PTP-KAPPA, receptor-type tyrosine-protein phosphatase kappa | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction