missing translation for 'onlineSavingsMsg'
Learn More

Pure Water Density Standard; UKAS ISO/IEC17025 and ISO 17034 certified, nominal density values: 0.9982 g/mL (20°C),0.9970 g/mL (25°C), 60mL, Paragon Scientific™

Product Code. 15625568
Change view
Click to view available options
Quantity:
60 mL
Unit Size:
60mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15625568 60 mL 60mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15625568 Supplier Paragon Scientific Supplier No. DENWAT
  • ONLINE EXCLUSIVE PRICE
    158.31 €
    151.20 € / 60mL
    Save 7.11 €
    4% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Paragon ISO 17025 / ISO 17034 pure water Density standard is designed for the verification of instruments used to measure density. Typically used for the verification of digital density meters. ISO 17025 / ISO 17034 certification under UKAS ensures full traceability to national standards.

  • Dual certified to ISO 17025 / ISO 17034 under our UKAS accreditation
  • Certified in accordance with primary level ASTM D1480 methodology
  • The reagent water meets the definition of Type II in the Specification ASTM D1193
  • Supplied in tamper evident packaging with a 12 month shelf life
  • Available as single bottle packs or triple pack kits
  • Manufactured in the United Kingdom

Specifications

Quantity 60 mL

Nominal values quoted. Certified Values will vary from batch to batch.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.