missing translation for 'onlineSavingsMsg'
Learn More

Keystone Cleanroom Products PureClean Self-Sealing 1421B Tyvek™ Autoclave Bag with Clear Film Side and Steam Indicator, 16 to 20 in. wide
GREENER_CHOICE

Product Code. 30042329 Shop All Keystone Cleanroom Products Products
Change view
Click to view available options
Size:
16 x 12
16 x 16
16 x 20
16 x 24
16 x 28
16 x 30
16 x 32
16 x 35
16 x 36
16 x 38
16 x 4
16 x 44
16 x 48
18 x 20
18 x 24
18 x 28
18 x 30
18 x 32
18 x 36
18 x 40
18 x 48
18 x 54
18 x 70
20 x 20
20 x 24
20 x 26
20 x 30
20 x 32
20 x 36
20 x 40
20 x 44
20 x 46
20 x 48
Unit Size:
Pack of 250
33 product options available for selection
Product selection table with 33 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
30042329 18 x 20 Pack of 250
30042323 20 x 48 Pack of 250
30042180 20 x 46 Pack of 250
30042213 20 x 44 Pack of 250
30042314 20 x 40 Pack of 250
30042306 20 x 36 Pack of 250
30042210 20 x 32 Pack of 250
30042318 20 x 30 Pack of 250
30042238 20 x 26 Pack of 250
30042200 20 x 24 Pack of 250
30042235 20 x 20 Pack of 250
30042227 18 x 70 Pack of 250
30042219 18 x 54 Pack of 250
30042214 18 x 48 Pack of 250
30042272 18 x 40 Pack of 250
30042293 18 x 36 Pack of 250
30042283 18 x 32 Pack of 250
30042254 18 x 30 Pack of 250
30042266 18 x 28 Pack of 250
30042295 18 x 24 Pack of 250
30042307 16 x 48 Pack of 250
30042310 16 x 44 Pack of 250
30042259 16 x 38 Pack of 250
30042288 16 x 36 Pack of 250
30042242 16 x 35 Pack of 250
30042234 16 x 32 Pack of 250
30042291 16 x 30 Pack of 250
30042316 16 x 28 Pack of 250
30042327 16 x 24 Pack of 250
30042218 16 x 20 Pack of 250
30042264 16 x 16 Pack of 250
30042282 16 x 12 Pack of 250
30042271 16 x 4 Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 33 options available.
33 options
This item is not returnable. View return policy
Product Code. 30042329 Supplier Keystone Cleanroom Products Supplier No. 6STKRP1820SS
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

PureClean™ Self-Sealing Autoclave Bags are made from a transparent film and 1421B DuPont™ Tyvek™, which is a new grade of uncoated, untreated, flexible Tyvek™ material specifically designed for pharmaceutical use in steam sterilization applications.

  • Uncoated Tyvek™ eliminates potential of residue deposits on equipment, post sterilization
  • Clear Side enables visual confirmation of dryness for sterilized parts and equipment, which is required by EU GMP Annex 1
  • Convenient Self-Seal Closure eliminates the need for autoclave tape or the validation and use of a heat sealer
  • Fast and consistent for equipment preparation area
  • Resists tearing and punctures
  • Superior microbial barrier
  • Manufactured and packaged in a certified ISO 6 cleanroom
  • Made in the USA
  • Certificates of Conformance sent with shipment

Specifications

Autoclavable Autoclavable
Certifications/Compliance COC
Closure Type Safe Seal
Color Clear, White
Disposable Single-use
For Use With (Application) Steam Sterilization of Pharmaceutical Equipment
Material Tyvek
Sterility Non-sterile
Latex Free Yes
Size 18 x 20
Temperature Range (Metric) 121 to 125 C
Thickness (English) 5.3 mil
Length (English) 20 in.
Width (English) 18 in.
Product Type Autoclave Bag
Length (Metric) 50.8 cm
Width (Metric) 45.7 cm
Thickness (Metric) 5.3 mil
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.