missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PureLink™ RNase A (20 mg/mL)
PureLink™ RNase A (ribonuclease A) is a bovine pancreatic ribonuclease that cleaves single-stranded RNA.
208.00 € - 375.00 €
Specifications
Concentration | 20 mg/mL |
---|---|
Packaging Type | Bottle |
Product Line | UltraPure™ |
Shipping Condition | Room Temperature |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10618703
|
Invitrogen™
12091021 |
10 mL |
208.00 €
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11508686
|
Invitrogen™
12091039 |
25 mL |
375.00 €
25mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- RNase A (ribonuclease A) is a bovine pancreatic ribonuclease that cleaves single-stranded RNA
- Used for the isolation of RNA-free DNA
Concentration
- 20 mg RNase A/mL in 50 mM Tris-HCl (pH 8.0), 10 mM EDTA
Specifications
20 mg/mL | |
UltraPure™ | |
Liquid |
Bottle | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.