missing translation for 'onlineSavingsMsg'
Learn More

Puromycin Aminonucleoside Hydrochloride, TRC

Product Code. 30410361
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30410361 10 mg 10mg
30417051 5 mg 5mg
30409361 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30410361 Supplier Toronto Research Chemicals Supplier No. TRCP84032110MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Puromycin Aminonucleoside Hydrochloride
Analyte or Component Names Puromycin Aminonucleoside Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C12H18N6O3 • HCl
Synonym 3'-Amino-3'-deoxy-N,N-dimethyladenosine Hydrochloride, 3'-Amino-3'-deoxy-N6,N6-dimethyladenosine Hydrochloride, 6-(Dimethylamino)-9-(3-amino-3-deoxy-β-D-ribofuranosyl)purine Hydrochloride, 6-Dimethylamino-9-(3-amino-3-deoxyribosyl)purine Hydrochloride, 9-(3-Amino-3-deoxy-β-D-ribofuranosyl)-6-(dimethylamino)-9H-purine Hydrochloride, Aminonucleoside Hydrochloride, Aminonucleoside Puromycin Hydrochloride, NSC 3056 Hydrochloride, PANS Hydrochloride, Aminonucleoside Puromycin Hydrochloride, SAN Hydrochloride, Stylomycin Aminonucleoside Hydrochloride
InChI Formula InChI=1S/C12H18N6O3.ClH/c1-17(2)10-8-11(15-4-14-10)18(5-16-8)12-9(20)7(13)6(3-19)21-12;/h4-7,9,12,19-20H,3,13H2,1-2H3;1H/t6-,7-,9-,12-;/m1./s1
Solution Type Neat
SMILES OC[C@@H]1[C@@H](N)[C@@H](O)[C@H](N2C(N=CN=C3N(C)C)=C3N=C2)O1.Cl
IUPAC Name (2R,3R,4S,5S)-4-amino-2-(6-(dimethylamino)-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3-ol hydrochloride
Industry Type Pharmaceutical
Molecular Weight (g/mol) 330.77
Quantity 10 mg
Formula Weight 330.12
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.