missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Puromycin aminonucleoside, MedChemExpress
Description
Puromycin aminonucleoside (NSC 3056) is the aminonucleoside portion of the antibiotic puromycin, and used in nephrosis animal models. Puromycin aminonucleoside induces apoptosis. Puromycin aminonucleoside is a reversible inhibitor of dipeptidyl peptidase II and cytosol alanyl aminopeptidase.
Specifications
Specifications
| Chemical Name or Material | Puromycin aminonucleoside |
| Physical Form | Solid |
| CAS | 58-60-6 |
| Color | Light Yellow |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C12H18N6O3 |
| For Use With (Application) | Cancer-Kinase/protease |
| Synonym | NSC 3056 |
| Solubility Information | H2O : 40 mg/mL (135.91 mM; ultrasonic and warming and heat to 50°C) ∣DMSO : 25 mg/mL (84.94 mM; ultrasonic and warming and heat to 60°C) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?