missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Puromycin (Dihydrochloride)

Product Code. 17100212
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17100212 50 mg 50mg
17110212 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17100212 Supplier STEMCELL Technologies Supplier No. 73342
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Antibiotic; Protein synthesis inhibitor

  • Puromycin is an aminonucleoside antibiotic, derived from Streptomyces alboniger, which acts as a protein synthesis inhibitor
  • It binds to the target ribosome site A, where it is transferred to the growing polypeptide chain causing premature chain termination (Lührmann et al.; Rodriguez-Fonseca et al.; Azzam & Algranati)
  • A biologically inactive form is generated when puromycin is N-acetylated by puromycin-N-acetyltransferase, allowing this gene to be used as a selective resistance marker (Vara et al.) 1985)
  • Selects for cells expressing puromycin-N-acetyltransferase resistance gene as a research tool (de la Luna & Ortín; Iwaki et al.)
  • Useful in CRISPR/Cas9 mammalian gene editing by selecting for successful Cas9-induced knock-in with puromycin resistance gene (Park et al.)
  • Possesses anti-tumor activity when tested against numerous cell lines (Foley & Eagle)

Specifications

CAS 58-58-2
Quantity 50 mg
Percent Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.