missing translation for 'onlineSavingsMsg'
Learn More

Melford Puromycin Dihydrochloride

Product Code. p-200087084
Click to view available options
Quantity:
25 mg
100 mg
500 mg
Unit Size:
100mg
25mg
500mg
This item is not returnable. View return policy

Product Code. 30018691

Brand: Melford P330200.1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puromycin Dihydrochloride is a potent antibiotic.

Puromycin Dihydrochloride is a potent antibiotic. It is an amino nucleoside antibiotic that mimics the 3'-end of aminoacyl-tRNA. It enters the ribosomal A site during translation, causing premature chain termination and inhibiting protein synthesis. Puromycin is incorporated into growing polypeptide chains, leading to the release of incomplete and nonfunctional peptides.

Widely used as a tool in cell biology to study protein synthesis and investigate translation processes. Its ability to terminate protein synthesis at the ribosome makes it valuable for various applications, including pulse-chase experiments and the analysis of nascent peptides. Puromycin resistance genes are often used as selectable markers in genetic engineering. Cells expressing the resistance gene can survive and proliferate in the presence of Puromycin, allowing for the selection of transfected or transduced cells.

Puromycin Dihydrochloride is a valuable tool for investigating protein synthesis, studying translation mechanisms, and selecting cells with specific genetic modifications.

Specifications

Chemical Name or Material Puromycin
CAS 58-58-2
Quantity 100 mg
Molecular Formula C22H29N7O5-2 HCl
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.