missing translation for 'onlineSavingsMsg'
Learn More

Puromycin dihydrochloride, MedChemExpress

Product Code. 16488718
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488718 10 mg 10mg
16444627 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16488718 Supplier Medchem Express Supplier No. HYB1743A
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Puromycin dihydrochloride (CL13900 dihydrochloride), an aminonucleoside antibiotic, inhibits protein synthesis.

Puromycin dihydrochloride (CL13900 dihydrochloride), an aminonucleoside antibiotic, inhibits protein synthesis.

Specifications

Chemical Name or Material Puromycin dihydrochloride
Physical Form Solid
CAS 58-58-2
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C22H31Cl2N7O5
For Use With (Application) COVID-19-immunoregulation
Synonym CL13900 dihydrochloride
Solubility Information H2O : 50 mg/mL (91.84 mM; Need ultrasonic and warming) ∣DMSO : 50 mg/mL (91.84 mM; Need ultrasonic) ∣Ethanol : 5 mg/mL (9.18 mM; ultrasonic and warming and heat to 60°C)
SMILES OC[C@@H]1[C@@H](NC([C@@H](N)CC2=CC=C(OC)C=C2)=O)[C@H]([C@H](N3C=NC4=C3N=CN=C4N(C)C)O1)O.[H]Cl.[H]Cl
Molecular Weight (g/mol) 544.43
Formula Weight 544.43
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.