missing translation for 'onlineSavingsMsg'
Learn More

TOKU-E™ Puromycin Dihydrochloride ReadyMade™ Solution

Product Code. 30044028
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 mL
5 x 1 mL
10 x 1 mL
Unit Size:
10mL
1mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30044028 5 x 1 mL 5mL
30044025 10 x 1 mL 10mL
30044013 20 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30044028 Supplier TOKU-E™ Supplier No. P0255x1ML
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puromycin Dihydrochloride ReadyMade™ Solution is provided as a sterile-filtered solution of Puromycin Dihydrochloride in 20 mM HEPES buffer.

  • CAS Number: 58-58-2
  • Molecular Formula: C22H29N7O5 · 2HCl
  • Molecular Weight: 544.43 g/mol
  • Storage Conditions: -20°C
  • Form: Liquid
  • Source: Streptomyces alboniger
  • Spectrum: Puromycin dihydrochloride is active against both prokaryotic and eukaryotic cells
  • Mechanism of Action: During translation, Puromycin enters the ribosomal 'A' site and disrupts peptide transfer. As a result, the ribosome stops and the peptide chain is terminated leading to a nonfunctional protein

Specifications

CAS 58-58-2
Certifications/Compliance Purity >98.0%
Chemical Name or Material Puromycin Dihydrochloride
Color Colorless to light yellow
Content And Storage -20°C
Description Puromycin Dihydrochloride ReadyMade™ Solution 10 mg/mL in HEPES buffer
Concentration 9.7 to 10.3 mg/mL
Form Clear, colorless to slight yellow solution
Product Type Antibiotic
Sterility Sterile
Sterilization Method Sterile-filtered
With Additives HEPES
Without Additives None
Formulation Liquid in HEPES Buffer (pH 6.2 to 6.8)
For Use With (Application) Eukaryotic Cell Culture Applications
pH 6.2 to 6.8
Product Line ReadyMade™ Solutions
Purity or Quality Grade >98%
Quantity 5 x 1 mL
Shipping Condition Dry Ice
Source Streptomyces alboniger
Species Bacteria
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.