missing translation for 'onlineSavingsMsg'
Learn More

TOKU-E™ Puromycin Dihydrochloride ReadyMade™ Solution

Product Code. 30044025
Encompass_Preferred
Click to view available options
Quantity:
20 mL
5 x 1 mL
10 x 1 mL
Unit Size:
10mL
1mL
5mL
This item is not returnable. View return policy

Product Code. 30044025

Brand: TOKU-E™ P02510x1ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puromycin Dihydrochloride ReadyMade™ Solution is provided as a sterile-filtered solution of Puromycin Dihydrochloride in 20 mM HEPES buffer.

  • CAS Number: 58-58-2
  • Molecular Formula: C22H29N7O5 · 2HCl
  • Molecular Weight: 544.43 g/mol
  • Storage Conditions: -20°C
  • Form: Liquid
  • Source: Streptomyces alboniger
  • Spectrum: Puromycin dihydrochloride is active against both prokaryotic and eukaryotic cells
  • Mechanism of Action: During translation, Puromycin enters the ribosomal 'A' site and disrupts peptide transfer. As a result, the ribosome stops and the peptide chain is terminated leading to a nonfunctional protein

Specifications

CAS 58-58-2
Certifications/Compliance Purity >98.0%
Chemical Name or Material Puromycin Dihydrochloride
Color Colorless to light yellow
Content And Storage -20°C
Description Puromycin Dihydrochloride ReadyMade™ Solution 10 mg/mL in HEPES buffer
Concentration 9.7 to 10.3 mg/mL
Form Clear, colorless to slight yellow solution
Product Type Antibiotic
Sterility Sterile
Sterilization Method Sterile-filtered
With Additives HEPES
Without Additives None
Formulation Liquid in HEPES Buffer (pH 6.2 to 6.8)
For Use With (Application) Eukaryotic Cell Culture Applications
pH 6.2 to 6.8
Product Line ReadyMade™ Solutions
Purity or Quality Grade >98%
Quantity 10 x 1 mL
Shipping Condition Dry Ice
Source Streptomyces alboniger
Species Bacteria
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.