missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PUS10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00 € - 550.00 €
Specifications
| Antigen | PUS10 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30229910
|
Novus Biologicals
NBP3-35626-20ul |
20 μL |
190.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30226591
|
Novus Biologicals
NBP3-35626-100ul |
100 μL |
550.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PUS10 Polyclonal antibody specifically detects PUS10 in Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| PUS10 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat | |
| CCDC139, coiled-coil domain containing 139, Coiled-coil domain-containing protein 139, DOBIMGC126729, EC 5.4.99, EC 5.4.99.-, FLJ32312, MGC126755, pseudouridine synthase 10, pseudouridylate synthase 10, Psi55 synthase, PUS1, putative tRNA pseudouridine synthase Pus10, tRNA pseudouridine 55 synthase, tRNA pseudouridylate synthase, tRNA-uridine isomerase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 290-529 of human PUS10 (NP_653310.2).,, Sequence:, VAGRYNKYSRNLPQTPWIIDGERKLESSVEELISDHLLAVFKAESFNFSSSGREDVDVRTLGNGRPFAIELVNPHRVHFTSQEIKELQQKINNSSNKIQVRDLQLVTREAIGHMKEGEEEKTKTYSALIWTNKAIQKKDIEFLNDIKDLKIDQKTPLRVLHRRPLAVRARVIHFMETQYVDEHHFRLHLKTQAGTYIKEFVHGDFGRTKPNIGSLMNVTADILELDVESVDVDWPPALDD | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:2000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 150962 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title