Learn More
Melford Penicillin G, Sodium Salt
Description
Penicillin G sodium is the sodium salt form of penicillin G, which is a narrow-spectrum beta-lactam antibiotic.
Penicillin G sodium interferes with bacterial cell wall synthesis. It inhibits the enzyme transpeptidase, which is involved in the cross-linking of peptidoglycan chains in the bacterial cell wall. This disruption weakens the cell wall structure, leading to cell lysis and death.
Also, Penicillin G is effective against a wide range of Gram-positive bacteria, including Streptococcus, Staphylococcus, and some anaerobic bacteria. It is less effective against Gram-negative bacteria.
Often used as a selective agent in culture media to isolate bacteria that are sensitive to this antibiotic. Its inclusion in culture medium allows for the specific growth of bacteria lacking resistance to penicillin G.
Specifications
Specifications
| Chemical Name or Material | Penicillin G, Sodium Salt |
| CAS | 69-57-8 |
| Quantity | 100 g |
| Molecular Formula | C16H17N2NaO4S |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.