missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PWAR1 Antibody (6A7H10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37677
This item is not returnable.
View return policy
Description
PWAR1 Monoclonal specifically detects PWAR1 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| PWAR1 | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 0.03% Sodium Azide | |
| Mouse | |
| Unpurified | |
| RUO | |
| 145624 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, ELISA | |
| 6A7H10 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000 | |
| D15S227E, PAR1, PAR-1, Prader Willi/Angelman Region 1, Prader Willi/Angelman Region RNA 1 | |
| Purified recombinant fragment of PAR1 expressed in E. Coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction