Learn More
PYCR2 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-17146-20UL
 View more versions of this product
Product Code. 18059425
  • 145.00 € / 20µL
Description
The PYCR2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PYCR2. This antibody recognizes human, mouse, rat antigen. The PYCR2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
PYCR2
1000 μg/mL
FLJ54750, P5C reductase 2, P5CR 2, P5CR2, PYCR2
20 μL
29920
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Immunofluorescence
CoraLite Plus 488
Rabbit
Primary
Human, Mouse, Rat
150 kDa