Learn More
PYGO1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA559927
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: RMPPHVPPRM SSPYCGPYSL RNQPHPFPQN PLGMGFNRPH AFNFGPHDNS SFGNPSYNNA LSQNVNMPNQ HFRQNPAENF SQIPPQNASQ VSNPDLASNF VPGNNSNFTS PLESNHSFIP PPNTFGQAKA PPPKQDFTQG ATK Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 88%.
PYGO1 is a protein coding gene involved in signal transduction through the Wnt pathway.Specifications
PYGO1 | |
Polyclonal | |
Unconjugated | |
PYGO1 | |
2600014C22Rik; PYGO1; pygopus 1; pygopus family PHD finger 1; pygopus homolog 1; pygopus-like protein 1; zgc:172214 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26108 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y3Y4 | |
PYGO1 | |
Recombinant protein corresponding to Human PYGO1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only