missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyr3, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
Description
Pyr3 is a selective inhibitor of transient receptor potential canonical channel 3 (TRPC3), with an IC50 of 700 nM for TRPC3-mediated Ca2+ influx.
Specifications
Specifications
| Chemical Name or Material | Pyr3 |
| Physical Form | Solid |
| CAS | 1160514-60-2 |
| Color | Off-White |
| Quantity | 25 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C16H11Cl3F3N3O3 |
| Solubility Information | DMSO : ≥ 125 mg/mL (273.74 mM) |
| SMILES | O=C(C1=C(C(F)(F)F)N(C2=CC=C(NC(/C(Cl)=C(Cl)/Cl)=O)C=C2)N=C1)OCC |
| Molecular Weight (g/mol) | 456.63 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction