missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyraflufen (free acid) 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Pyraflufen monohydrate (free acid) |
| CAS | 129630-17-7 |
| Analyte or Component Names | Pyraflufen monohydrate (free acid) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H9 Cl2 F3 N2 O4 |
| Concentration | 100 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | Acetic acid, [2-chloro-5-[4-chloro-5-(difluoromethoxy)-1-methyl-1H-pyrazol-3-yl]-4-fluorophenoxy]- (9CI), 2-[2-Chloro-5-[4-chloro-5-(difluoromethoxy)-1-methyl-1H-pyrazol-3-yl]-4-fluorophenoxy]acetic acid, 2-Chloro-5-(4-chloro-5-difluoromethoxy-1-methylpyrazol-3-yl)-4-fluorophenoxyacetic acid, E 1, Pyraflufen |
| InChI Formula | InChI=1S/C13H9Cl2F3N2O4/c1-20-12(24-13(17)18)10(15)11(19-20)5-2-8(23-4-9(21)22)6(14)3-7(5)16/h2-3,13H,4H2,1H3,(H,21,22) |
| SMILES | Cn1nc(c(Cl)c1OC(F)F)c2cc(OCC(=O)O)c(Cl)cc2F |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?