missing translation for 'onlineSavingsMsg'
Learn More

Pyrazinamide, MedChemExpress

Product Code. 16498438
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498438 10 g 10g
16484317 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16498438 Supplier Medchem Express Supplier No. HYB0271
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Pyrazinamide (Pyrazinecarboxamide; Pyrazinoic acid amide) is a potent and orally active antitubercular antibiotic. Pyrazinamide is a prodrug that is converted to the active form pyrazinoic acid (POA) by PZase/nicotinamidase encoded by the pncA gene in M. tuberculosis.

Pyrazinamide (Pyrazinecarboxamide

Specifications

Chemical Name or Material Pyrazinamide
Physical Form Solid
CAS 98-96-4
Color White
Quantity 10 g
Purity Grade Notes Research
Molecular Formula C5H5N3O
For Use With (Application) COVID-19-immunoregulation
Synonym Pyrazinecarboxamide Pyrazinoic acid amide
Solubility Information DMSO : ≥ 50 mg/mL (406.14 mM) ∣H2O : 6.67 mg/mL (54.18 mM; Need ultrasonic)
SMILES O=C(C1=NC=CN=C1)N
Molecular Weight (g/mol) 123.11
Formula Weight 123.11
Percent Purity 99.95%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.