missing translation for 'onlineSavingsMsg'
Learn More

Pyrazinamide, TRC

Product Code. 17955085
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17955085 10 g 10g
17921385 25 g 25g
17914185 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17955085 Supplier Toronto Research Chemicals Supplier No. TRCP84060010G
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-96-4
Molecular Formula C5 H5 N3 O
Molecular Weight (g/mol) 123.11
Synonym Pyrazinamide, 2-Pyrazinecarboxamide, Pyrazinecarboxamide (8CI,9CI), 2-(Aminocarbonyl)pyrazine, 2-Carbamylpyrazine, 2-Carboxamidopyrazine, Aldinamid, Aldinamide, D 50, Eprazin, Farmizina, Isopas, MK 56, NSC 14911, Novamid, Pirazimida, Pirazinamid, Pyrafat, Pyramizade, Pyrazide, Pyrazinamide, Pyrazine carboxylamide, Pyrazinecarboxylic acid amide, Pyrazinoic acid amide, Rifafour e-200, Rifcin, Rozide, Tebrazid, Unipyranamide, Zinamide, α-Pyrazinamide
IUPAC Name pyrazine-2-carboxamide
SMILES NC(=O)c1cnccn1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C5H5N3O/c6-5(9)4-3-7-1-2-8-4/h1-3H,(H2,6,9)
Formula Weight 123.0433
Chemical Name or Material Pyrazinamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.