accessibility menu, dialog, popup

UserName

H2NONN

CAS RN 98-96-4

IUPAC Name: pyrazine-2-carboxamide
Synonyms: pyrazinamide pyramide eprazin zinamide pirilene pyrafat lynamide pyrazinamdie corsazinmid aldinamid
Molecular Weight (g/mol): 123.12
Molecular Formula: C5H5N3O
InChi Key: IPEHBUMCGVEMRF-UHFFFAOYSA-N
SMILES: NC(=O)C1=CN=CC=N1
Molecular Weight (g/mol) 
  • (10)
  • (4)
Quantity 
  • (1)
  • (4)
  • (2)
  • (1)
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (1)
  • (3)
  • (2)
Physical Form 
  • (3)
  • (3)
  • (2)
Color 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (2)
  • (10)
  • (4)
  • (1)
  • (4)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)