missing translation for 'onlineSavingsMsg'
Learn More

Pyrazine-2,5-dicarboxylic acid, 95%

Product Code. 15452159
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15452159 1 g 1g
15472159 250 mg 250mg
15462159 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15452159 Supplier Thermo Scientific Alfa Aesar Supplier No. H64795.03
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 122-05-4 | C6H4N2O4 | 168.11 g/mol

It is an pyrazine used for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an pyrazine used for proteomics research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-05-4
Molecular Formula C6H4N2O4
Molecular Weight (g/mol) 168.11
MDL Number MFCD00216919
InChI Key GMIOYJQLNFNGPR-UHFFFAOYSA-N
Synonym 2,5-pyrazinedicarboxylic acid, 2,5-pyrazinedicarboxylicacid, pubchem8868, 2,5-pyrazine-diacid, ksc177q1r, pyrazinedicarboxylic-2,5 acid
PubChem CID 255280
IUPAC Name pyrazine-2,5-dicarboxylic acid
SMILES OC(=O)C1=CN=C(C=N1)C(O)=O

Specifications

Melting Point 255°C to 260°C (literature)
Quantity 1 g
Beilstein 139985
Solubility Information Slightly soluble in water.
Formula Weight 168.11
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Pyrazine-2,5-dicarboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.