missing translation for 'onlineSavingsMsg'
Learn More

Pyrene, 98%

Product Code. 10673132 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10673132 25 g Glass bottle 25g
10245101 100 g Glass bottle 100g
10465755 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10673132 Supplier Thermo Scientific Supplier No. 180830250
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 129-00-0 | C16H10 | 202.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129-00-0
Molecular Formula C16H10
Molecular Weight (g/mol) 202.25
MDL Number MFCD00004136
InChI Key BBEAQIROQSPTKN-UHFFFAOYSA-N
Synonym benzo def phenanthrene, pyren, beta-pyrene, .beta.-pyrene, pyren german, unii-9e0t7wfw93, ccris 1256, pyrene def phenanthrene, coal tar pitch volatiles:pyrene
PubChem CID 31423
ChEBI CHEBI:39106
IUPAC Name pyrene
SMILES C1=CC2=C3C(=C1)C=CC4=CC=CC(=C43)C=C2

Specifications

Melting Point 148°C to 152°C
Color Green-Yellow to Yellow
Boiling Point 393°C
Flash Point 210°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 25 g
Beilstein 05,693
Fieser 04,414
Merck Index 15,8074
Solubility Information Solubility in water: almost insoluble. Other solubilities: soluble in ethanol,ether,benzene and toluene,slightly soluble in carbon tetrachloride
Formula Weight 202.25
Percent Purity 98%
Physical Form Crystals and/or Chunks
Chemical Name or Material Pyrene
Show More Show Less
compliance-icons
Product Identifier
  • Pyrene
Signal Word
  • Warning
Hazard Category
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.