missing translation for 'onlineSavingsMsg'
Learn More

Pyrene, 98%

Product Code. 11328015
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11328015 100 g 100g
11318015 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11328015 Supplier Thermo Scientific Alfa Aesar Supplier No. L08162.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 129-00-0 | C16H10 | 202.256 g/mol

Pyrene and its derivatives are used commercially to make dyes and dye precursors, for example pyranine and naphthalene-1,4,5,8-tetracarboxylic acid. It is used as a probe to determine solvent environments and fluorescence spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyrene and its derivatives are used commercially to make dyes and dye precursors, for example pyranine and naphthalene-1,4,5,8-tetracarboxylic acid. It is used as a probe to determine solvent environments and fluorescence spectroscopy.

Solubility
Soluble in toluene. Insoluble in water.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129-00-0
Molecular Formula C16H10
Molecular Weight (g/mol) 202.256
MDL Number MFCD00004136
InChI Key BBEAQIROQSPTKN-UHFFFAOYSA-N
Synonym benzo def phenanthrene, pyren, beta-pyrene, .beta.-pyrene, pyren german, unii-9e0t7wfw93, ccris 1256, pyrene def phenanthrene, coal tar pitch volatiles:pyrene
PubChem CID 31423
ChEBI CHEBI:39106
IUPAC Name pyrene
SMILES C1=CC2=C3C(=C1)C=CC4=CC=CC(=C43)C=C2

Specifications

Melting Point 148°C to 152°C
Density 1.271
Boiling Point 393°C
Flash Point 208°C (406°F)
Quantity 100 g
Beilstein 1307225
Merck Index 14,7963
Solubility Information Soluble in toluene. Insoluble in water.
Formula Weight 202.26
Percent Purity 98%
Chemical Name or Material Pyrene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.