missing translation for 'onlineSavingsMsg'
Learn More

Pyrene, TRC

Product Code. 30136958
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136958 10 g 10g
30136669 25 g 25g
18000430 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136958 Supplier Toronto Research Chemicals Supplier No. TRCP84900010G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129-00-0
Molecular Formula C16 H10
Molecular Weight (g/mol) 202.25
Synonym Pyrene, Benzo[def]phenanthrene, NSC 17534, NSC 66449
IUPAC Name pyrene
SMILES c1cc2ccc3cccc4ccc(c1)c2c34

Specifications

Quantity 10 g
InChI Formula InChI=1S/C16H10/c1-3-11-7-9-13-5-2-6-14-10-8-12(4-1)15(11)16(13)14/h1-10H
Formula Weight 202.0783
Percent Purity >95%
Chemical Name or Material Pyrene
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.