missing translation for 'onlineSavingsMsg'
Learn More

Pyrene, TRC

Product Code. 18000430
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18000430 1 g 1g
30136958 10 g 10g
30136669 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18000430 Supplier Toronto Research Chemicals Supplier No. TRCP8490001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129-00-0
Formula Weight 202.0783
InChI Formula InChI=1S/C16H10/c1-3-11-7-9-13-5-2-6-14-10-8-12(4-1)15(11)16(13)14/h1-10H
IUPAC Name pyrene
Molecular Formula C16 H10
Molecular Weight (g/mol) 202.25
Percent Purity >95%
SMILES c1cc2ccc3cccc4ccc(c1)c2c34
Synonym Pyrene, Benzo[def]phenanthrene, NSC 17534, NSC 66449
Chemical Name or Material Pyrene
Grade HPLC
Show More Show Less

Specifications

Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.