missing translation for 'onlineSavingsMsg'
Learn More

Pyrethrins (technical), Dr. Ehrenstorfer

Product Code. 30310900
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30310900 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30310900 Supplier Dr. Ehrenstorfer Supplier No. DRECA16620000
Big Savings for Better Separation
Use promo code 27992
View all promotions
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pyrethrins
CAS 8003-34-7
Analyte or Component Names Pyrethrins
Shipping Condition Ice pack (-20°C)
Certifications/Compliance ISO/IEC 17025
Synonym Pyrethrins, Pyrethrins and Pyrethroids, Biopiren plus, Chrysanthemates pyrethrins and pyrethroids, Diatect, Firmotox, Goldgeist forte, Nettafid, Parexan, Piresan, Pyganic, Pyganic MUP 20, Pyre-sin Plus, Pyrellin, Pyrethrine, Pyrethrines, Pyrethrins and Pyrethroids, chrysanthemates, Pyrethrins and Pyrethroids, esters, Pyrethrins, chrysanthemates, Pyrethro Vioryl 5SC, Pyrethrum, Pyrethrum (insecticide), Pyrethrum FS, Pyrevert, Pyrocide 50, Spruzit, Take Down, Take Down Garden Spray
SMILES C/C(C)=C/C1C(C)(C)[C@@H]1C(O[C@@H]2C(C)=C(C/C=C/C=C)C(C2)=O)=O.C/C(C([CH3]=O)=O)=C\C3C(C)(C)[C@@H]3C(OC4C(C)=C(C/C=C/C=C)C(C4)=O)=O
Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.