missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-2,6-dicarboxylic acid, 98%

Product Code. 11444973
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11444973 100 g 100g
11434973 25 g 25g
11454973 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11444973 Supplier Thermo Scientific Alfa Aesar Supplier No. A12263.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 499-83-2 | C7H5NO4 | 167.12 g/mol

Pyridine-2,6-dicarboxylic acid is used in the preparation of dipicolinato ligated lanthanide and transition metal complexes. It acts as a chelating agent for chromium, zinc, manganese, copper, iron and molybdenum. Its calcium-dipcolinic acid complex is used to protect deoxyribonucleic acid (DNA) from heat denaturation which enhances the DNA stability. It plays an important role as a marker for the effectiveness of sterilization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyridine-2,6-dicarboxylic acid is used in the preparation of dipicolinato ligated lanthanide and transition metal complexes. It acts as a chelating agent for chromium, zinc, manganese, copper, iron and molybdenum. Its calcium-dipcolinic acid complex is used to protect deoxyribonucleic acid (DNA) from heat denaturation which enhances the DNA stability. It plays an important role as a marker for the effectiveness of sterilization.

Solubility
Insoluble in water.

Notes
Incompatible with acids, bases and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 499-83-2
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00006299
InChI Key WJJMNDUMQPNECX-UHFFFAOYSA-N
Synonym 2,6-pyridinedicarboxylic acid, dipicolinic acid, 2,6-dipicolinic acid, dipicolinate, 2,6-dicarboxypyridine, 2,6-pyridinedicarboxylate, unii-ue81s5cq0g, 2,6-pyridinedicarboxylicacid, ue81s5cq0g, 2,6-pyridinedicarboxylic acid dipicolinic acid
PubChem CID 10367
ChEBI CHEBI:46837
IUPAC Name pyridine-2,6-dicarboxylic acid
SMILES C1=CC(=NC(=C1)C(=O)O)C(=O)O

Specifications

Melting Point ∼245°C (decomposition)
Flash Point 188°C (370°F)
Quantity 100 g
Beilstein 131629
Solubility Information Insoluble in water.
Formula Weight 167.12
Percent Purity 98%
Chemical Name or Material Pyridine-2,6-dicarboxylic acid
compliance-icons
Product Identifier
  • Pyridine-2,6-dicarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 2,6-Pyridinedicarboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.