missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-3-boronic acid 1,3-propanediol ester, 98+%

Product Code. p-7046975
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11393896

Brand: Thermo Scientific Alfa Aesar L17010.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 131534-65-1 | C8H10BNO2 | 162.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131534-65-1
Molecular Formula C8H10BNO2
Molecular Weight (g/mol) 162.98
MDL Number MFCD01075749
InChI Key ZUUZNOFTKGDLLN-UHFFFAOYSA-N
Synonym 3-1,3,2-dioxaborinan-2-yl pyridine, 3-pyridineboronic acid 1,3-propanediol ester, pyridine-3-boronic acid 1,3-propanediol cyclic ester, 1,3-propanediol cyclic ester, 2-3-pyridyl-1,3,2-dioxaborinane, 3-1,3,2 dioxaborinan-2-yl-pyridine, pyridine-3-boronic acid 1,3-propanediol ester, 3-pyridineboronic acid glycol ester, 2-3-pyridyl-1,3,2-dioxaborane, 2-pyridin-3-yl-1,3,2-dioxaborinane
PubChem CID 4198005
IUPAC Name 3-(1,3,2-dioxaborinan-2-yl)pyridine
SMILES C1COB(OC1)C1=CC=CN=C1

Specifications

Melting Point 100°C to 104°C
Quantity 1 g
Beilstein 6138748
Formula Weight 162.98
Percent Purity ≥98%
Chemical Name or Material Pyridine-3-boronic acid 1,3-propanediol ester

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.