missing translation for 'onlineSavingsMsg'
Learn More

Pyridine-4-boronic acid hydrochloride, 95%

Product Code. 15479397
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15479397 1 g 1g
15489397 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15479397 Supplier Thermo Scientific Alfa Aesar Supplier No. H52648.03
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 913835-65-1 | C5H7BClNO2 | 159.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 913835-65-1
Molecular Formula C5H7BClNO2
Molecular Weight (g/mol) 159.38
MDL Number MFCD06201888
InChI Key NCZXYQJUECNKAD-UHFFFAOYSA-N
Synonym pyridine-4-boronic acid hydrochloride, pyridin-4-ylboronic acid hydrochloride, pyridine-4-boronic acid, hcl, 4-pyridineboronic acid, hydrochloride, pyridine-4-boronic acid hcl, pyridine-4-boronic acid-hydrogen chloride, boronicacid, b-4-pyridinyl-, hydrochloride 1:1, c5h6bno2.hcl, pyridineboronic acid, 4-, hcl, 4-pyridineboronic acid, hcl
PubChem CID 44118740
IUPAC Name pyridin-4-ylboronic acid;hydrochloride
SMILES Cl.OB(O)C1=CC=NC=C1

Specifications

Melting Point 340°C
Quantity 1 g
Formula Weight 159.38
Percent Purity 95%
Chemical Name or Material Pyridine-4-boronic acid hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.