missing translation for 'onlineSavingsMsg'
Learn More

Pyridine hydrobromide, 98%

Product Code. 11409576
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11409576 50 g 50g
11419576 250 g 250g
11499566 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11409576 Supplier Thermo Scientific Alfa Aesar Supplier No. A19314.18
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 18820-82-1 | C5H6BrN | 160.014 g/mol

Pyridine hydrobromide may be used in chemical synthesis studies. Pyridine hydrobromide is quite useful for brominations of ketones on a micro or semimicro scale.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyridine hydrobromide may be used in chemical synthesis studies. Pyridine hydrobromide is quite useful for brominations of ketones on a micro or semimicro scale.

Solubility
Soluble in water.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18820-82-1
Molecular Formula C5H6BrN
Molecular Weight (g/mol) 160.014
MDL Number MFCD00012801
InChI Key BBFCIBZLAVOLCF-UHFFFAOYSA-N
Synonym pyridine hydrobromide, pyridinium bromide, pyridine, hydrobromide, pyridine, hydrobromide 1:1, pyridine hbr, pyridinehydrobromide, acmc-1bovh, ksc178o2t, pyridine hydrobromide, purum at
PubChem CID 87810
IUPAC Name pyridine;hydrobromide
SMILES C1=CC=NC=C1.Br

Specifications

Melting Point 216°C to 220°C
Quantity 50 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 160.01
Percent Purity 98%
Chemical Name or Material Pyridine hydrobromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.