missing translation for 'onlineSavingsMsg'
Learn More

Pyrimitate, TRC

Product Code. 30137718
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137718 500 mg 500mg
30132610 250 mg 250mg
30011537 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137718 Supplier Toronto Research Chemicals Supplier No. TRCP997785500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5221-49-8
Molecular Formula C11 H20 N3 O3 P S
Molecular Weight (g/mol) 305.33
Synonym 4-Pyrimidinol, 2-(dimethylamino)-6-methyl-, O-ester with O,O-diethyl phosphorothioate (8CI), Diothyl, ICI 29661, O,O-Diethyl O-(2-dimethylamino-4-methyl-6-pyrimidyl) phosphorothioate, O,O-Diethyl O-(2-dimethylamino-4-methylpyrimidin-6-yl)phosphorothionate, O,O-Diethyl O-[2-dimethylamino-6-methyl-4-pyrimidinyl] phosphorothioate, Pyrimital, Pyrimitate, Pyrimithate, Phosphorothioic acid, O-[2-(dimethylamino)-6-methyl-4-pyrimidinyl] O,O-diethyl ester
IUPAC Name 4-diethoxyphosphinothioyloxy-N,N,6-trimethylpyrimidin-2-amine
SMILES CCOP(=S)(OCC)Oc1cc(C)nc(n1)N(C)C

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C11H20N3O3PS/c1-6-15-18(19,16-7-2)17-10-8-9(3)12-11(13-10)14(4)5/h8H,6-7H2,1-5H3
Formula Weight 305.0963
Percent Purity >95%
Chemical Name or Material Pyrimitate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.