missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PYROXD1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
415.00 € - 572.00 €
Specifications
| Antigen | PYROXD1 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18151929
|
Novus Biologicals
NBP2-38404 |
0.1 mL |
572.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18630186
|
Novus Biologicals
NBP2-38404-25ul |
25 μL |
415.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PYROXD1 Polyclonal specifically detects PYROXD1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| PYROXD1 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q8WU10 | |
| 79912 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: MCEVKKIYLQDEFRILKKKSFTFPRDHKSVTADTEMWPVYVELTNEKIYGCDFIVSATGVTPNVEPFLHGNSFDLGEDGGLKVDDHMHTSLPDIYAAG | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| DKFZp762G094, EC 1.8.1, EC 1.8.1.-, FLJ22028, pyridine nucleotide-disulfide oxidoreductase domain-containing protein 1, pyridine nucleotide-disulphide oxidoreductase domain 1 | |
| PYROXD1 | |
| IgG | |
| Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title