missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Q-VD-OPH Inhibitor Peptide

Product Code. 18748003 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
3 mg
5 mg
Unit Size:
1mg
3mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18748003 1 mg 1mg
18110683 3 mg 3mg
18180784 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18748003 Supplier R&D Systems™ Supplier No. NBP229391
  • ONLINE EXCLUSIVE PRICE
    480.86 €
    396.90 € / 1mg
    Save 83.96 €
    17% Off
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Applications: Functional, In vitro assay

TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Quantity 1 mg
Product Line Novus

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.