Learn More
BINDER™ Qualification Documents IQ/OQ /PQ
Product Code. 16464009 Shop All BINDER Products
Click to view available options
For Use With (Equipment):
BINDER Chambers (Series BD, BF, ED, FD, FED, FDL, FP, M, C, CB, KB, KT, MK, MKT, VD, VDL, UF V)
BINDER Chambers (Series KBF P, KBF LQC, KBW, KBWF)
BINDER Chambers (Series KBF, KBF-S, KMF, MKF, MKFT)
Product Type:
IQ/OQ Documentation (Hardcopy)
IQ/OQ Documentation (PDF)
Unit Size:
Each
Product Code. 16464009
Brand: BINDER™ 70070006
Qualification documents - IQ/OQ/PQ documents – supporting documents for validation performed by customers, according to customer requirements, PQ section added to qualification folder IQ/OQ; parameters: temperature, CO2, O2 – or pressure, depending on unit.
  • Parameters depending on unit
  • Available as hardcopy inside folder or PDF document
Specifications
For Use With (Equipment) BINDER Chambers (Series KBF, KBF-S, KMF, MKF, MKFT)
Description Qualification documents - Hard copy inside folder, IQ/OQ/PQ documents - supporting documents for validation performed by customers, according to customer requirements, PQ section added to qualification folder IQ/OQ; parameters: temperature and humidity values
Duration On request
Product Type IQ/OQ Documentation (Hardcopy)
Service Offering IQ/OQ/PQ with Completion of Documents; Parameters: Temperature and Humidity Values