Learn More
Socorex™ Qualitix™ Sterilization Paper Tray
Advantageously replacing stainless steel trays, Qualitix™ Sterilization paper tray are fully autoclavable by either EO, Steam or Gamma Ray.
Brand:  Socorex™ 3220.1526
Additional Details : Weight : 0.75000kg
Product Code. 17296800
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Rigid medical PE coated paper material
  • Advantageously replacing stainless steel trays
  • One size fits most devices
  • Suitable for EO, steam and gamma sterilization
  • Fit for use as such, or in combination with Qualitix™ sterilization pouches
  • Conformity with ISO 14971, ISO 11135, ISO 17665-06, CE standard
Specifications
Specifications
Sterilization tray
ISO 14971, ISO 11135, ISO 17665-06, CE standard
Single-use
Autoclave or EO or Gamma Rays
Polyethylene-coated Medical Paper
500
150 x 265 x 15 mm
265 mm
150 mm
15 mm
Autoclavable
White
Sterilization
Paper tray only
Non-sterile
50
10.5 in.
6 in.
0.6 in.