missing translation for 'onlineSavingsMsg'
Learn More

Quality Control Standards 23 For ICP, SPEX CertiPrep™

Product Code. 11475938 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
125 mL
Unit Size:
125mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11475938 125 mL 125mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 11475938 Supplier SPEX Certiprep Supplier No. QC23
  • ONLINE EXCLUSIVE PRICE
    721.01 €
    654.56 € / 125mL
    Save 66.45 €
    9% Off
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ag1000μg/mL
Al1000μg/mL
B1000μg/mL
Ba1000μg/mL
Bi1000μg/mL
Ca1000μg/mL
Cd1000μg/mL
Co1000μg/mL
Cr1000μg/mL
Cu1000μg/mL
Fe1000μg/mL
Ga1000μg/mL
In1000μg/mL
K1000μg/mL
Li1000μg/mL
Mg1000μg/mL
Mn1000μg/mL
Na1000μg/mL
Ni1000μg/mL
Pb1000μg/mL
Sr1000μg/mL
Tl1000μg/mL
Zn1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material Quality Control Standard 23
Name Note Standard 23
Packaging Triple-leached LDPE bottle
CAS 7697-37-2, 74-97-5, 7439-96-5, 7439-92-1, 7440-50-8
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
CAS Max % 10.0, 90.0
Analyte or Component Names Ag,Al,B
Technique AA, ICP
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
Element Ag, Al, B
Solvent or Matrix 10% HNO3
Quantity 125 mL
Grade Assurance
Show More Show Less

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.