missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ QuantStudio™ Absolute Q™ MAP16 Digital PCR Gasket Strip
Shop All Applied Biosystems Products
Click to view available options
Quantity:
60 Pk.
Description
QuantStudio Absolute Q MAP16 gasket strip is one component of the QuantStudio Absolute Q MAP16 Plate Kit (Cat. No. A52865). The gasket strip is intended to supplement the materials supplied in the plate kit for those customers who require additional materials. The gasket strips help with sealing the plate, maintaining pressure, and ensuring thermal uniformity.
Specifications
Specifications
| Barcode | No Barcode |
| Color | Blue |
| Product Type | dPCR Gasket |
| For Use With (Equipment) | MAP16 dPCR Plate |
| Sterility | Sterile |
| Quantity | 60 Pk. |
For Research Use Only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction