missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ QuantStudio™ Absolute Q™ MAP16 Plate Kit

Product Code. 17407823 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
12 Plates
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17407823 12 Plates Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17407823 Supplier Applied Biosystems™ Supplier No. A52865
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Applied Biosystems QuantStudio Absolute Q MAP16 Plate Kit is a key component of the QuantStudio Absolute Q Digital PCR System.

The Applied Biosystems QuantStudio Absolute Q MAP16 Plate Kit is a key component of the QuantStudio Absolute Q Digital PCR System. The system is powered by the patented microfluidic array plate (MAP) technology that enables consistent and automated separation of the sample into 20,000 microchambers with industry leading <1% coefficient of variation (CV). The QuantStudio Absolute Q MAP16 consumable uses microinjection-molded plates to overcome common challenges of inconsistency and high dead volume. This novel approach provides many benefits including: precise microchamber volume, consistent sample distribution, and greater than 98% of loaded volume analyzed.

The unparalleled precision and accuracy of the Absolute Q system provides confidence in measurements across replicates and experiments.

TRUSTED_SUSTAINABILITY

Specifications

Barcode Includes Barcode
Content And Storage  Includes MAP16 plates, gaskets, and Absolute Q Isolation Buffer (Cat. No. A52730)
For Use With (Equipment) QuantStudioTM Absolute QTM Digital PCR System
No. of Wells 16
For Use With (Application) Digital PCR
No. per Case 12 plates
No. per Pack 12 plates
Quantity 12 Plates
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.