missing translation for 'onlineSavingsMsg'
Learn More

Quercetin dihydrate, 97%

Product Code. 11402478
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11402478 100 g 100g
11492468 25 g 25g
11412478 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11402478 Supplier Thermo Scientific Alfa Aesar Supplier No. A15807.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6151-25-3 | C15H14O9 | 338.27 g/mol

Quercetin dihydrate used as a Inhibitor of protein tyrosine kinase, mitochondrial ATPase and phosphodiesterase inhibitor. It Inhibits PI3-kinase activity and slightly inhibits PIP kinase activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Quercetin dihydrate used as a Inhibitor of protein tyrosine kinase, mitochondrial ATPase and phosphodiesterase inhibitor. It Inhibits PI3-kinase activity and slightly inhibits PIP kinase activity.

Solubility
Soluble in DMSO (68 mg/ml at 25°C), Acetic Acid (1 mg/ml), Aqueous Alkali (1 mg/ml), water (<1 at 25°C), and ethanol (21 mg/ml at 25°C).

Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6151-25-3
Molecular Formula C15H14O9
Molecular Weight (g/mol) 338.27
MDL Number MFCD00149487
InChI Key GMGIWEZSKCNYSW-UHFFFAOYSA-N
Synonym quercetin dihydrate, 2-3,4-dihydroxyphenyl-3,5,7-trihydroxy-4h-chromen-4-one dihydrate, quercetine dihydrate, quercetin, dihydrate, ccris 3304, 3,3',4',5,7-pentahydroxyflavone dihydrate, quercetin dihydrate sophoretin, flavone, 3,3',4',5,7-pentahydroxy-, dihydrate, dsstox_cid_1219, 2-3,4-dihydroxyphenyl-3,5,7-trihydroxy-4h-1-benzopyran-4-one dihydrate
PubChem CID 5284452
IUPAC Name 2-(3,4-dihydroxyphenyl)-3,5,7-trihydroxychromen-4-one;dihydrate
SMILES O.O.OC1=CC(O)=C2C(OC(=C(O)C2=O)C2=CC=C(O)C(O)=C2)=C1

Specifications

Melting Point 310°C to 315°C
Assay Percent Range 97%
Odor Odorless
Quantity 100 g
UN Number UN2811
Beilstein 317313
Merck Index 14,8034
Solubility Information Soluble in DMSO (68mg/ml at 25°C),Acetic Acid (1mg/ml),Aqueous Alkali (1mg/ml),water (<1 at 25°C),and ethanol (21mg/ml at 25°C).
Formula Weight 338.28 (302.25 Anhydrous)
Percent Purity 97%
Chemical Name or Material Quercetin dihydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Quercetin dihydrate
Signal Word
  • Danger||Warning
Hazard Category
  • Acute toxicity Category 3
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H341-Suspected of causing genetic defects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Quercetin dihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.