missing translation for 'onlineSavingsMsg'
Learn More

Quinine, MedChemExpress

Product Code. 16472409
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16472409 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16472409 Supplier Medchem Express Supplier No. HYD0143
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Quinine is an alkaloid derived from the bark of the cinchona tree, acts as an anti-malaria agent. Quinine is a potassium channel inhibitor that inhibits WT mouse Slo3 (KCa5.1) channel currents evoked by voltage pulses to +100 mV with an IC50 of 169 μM.

Quinine is an alkaloid derived from the bark of the cinchona tree, acts as an anti-malaria agent. Quinine is a potassium channel inhibitor that inhibits WT mouse Slo3 (KCa5.1) channel currents evoked by voltage pulses to +100 mV with an IC50 of 169 μM.

Specifications

Chemical Name or Material Quinine
Physical Form Solid
CAS 130-95-0
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C20H24N2O2
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 100 mg/mL (308.24 mM) ∣H2O : 0.1 mg/mL (0.31 mM; Need ultrasonic)
SMILES O[C@H](C1=CC=NC2=C1C=C(OC)C=C2)[C@@]3([H])[N@](C[C@@H]4C=C)CC[C@H]4C3
Molecular Weight (g/mol) 324.42
Formula Weight 324.42
Percent Purity 97.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.