missing translation for 'onlineSavingsMsg'
Learn More

Quinoline-8-carboxaldehyde, 97%

Product Code. 11335445
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. Quantity unitSize
11335445 1 g 1g
11355445 250 mg 250mg
11345445 5 g 5g
3 options
This item is not returnable. View return policy

Product Code. 11335445

Brand: Thermo Scientific Alfa Aesar L19642.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 38707-70-9 | C10H7NO | 157.172 g/mol

Quinoline-8-carboxaldehyde is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Quinoline-8-carboxaldehyde is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Insoluble in water.

Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38707-70-9
Molecular Formula C10H7NO
Molecular Weight (g/mol) 157.172
MDL Number MFCD00805837
InChI Key OVZQVGZERAFSPI-UHFFFAOYSA-N
Synonym 8-quinolinecarboxaldehyde, 8-quinolinecarbaldehyde, quinoline-8-carboxaldehyde, 8-quinoline carbaldehyde, 8-formylquinoline, ccris 6968, timtec-bb sbb010129, 8-quinolinecarboaldehyde, acmc-209j1k, ksc224g8j
PubChem CID 170103
IUPAC Name quinoline-8-carbaldehyde
SMILES C1=CC2=C(C(=C1)C=O)N=CC=C2

Specifications

Melting Point 92°C to 94°C
Quantity 1 g
Beilstein 113039
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 157.17
Percent Purity ≥98%
Chemical Name or Material Quinoline-8-carboxaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.